missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydro-1,4-benzodioxine-5-carbaldehyde, Thermo Scientific™
£54.15 - £443.00
Chemical Identifiers
CAS | 29668-43-7 |
---|---|
Molecular Formula | C9H8O3 |
Molecular Weight (g/mol) | 164.16 |
MDL Number | MFCD00239451 |
InChI Key | BJXUCBAQZJITKD-UHFFFAOYSA-N |
Synonym | 2,3-dihydrobenzo b 1,4 dioxine-5-carbaldehyde, 2,3-dihydro-benzo 1,4 dioxine-5-carbaldehyde, 1,4-benzodioxin-5-carboxaldehyde, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxine-5-carbaldehyd, 2,3-dihydro-1,4-benzodioxin-5-carboxaldehyde, 2,3-ethylenedioxybenzaldehyde, 2,3-dihydrobenzodioxin-5-carboxaldehyde, 1,4-benzodioxane-5-carboxaldehyde, 2h,3h-benzo e 1,4-dioxin-5-carbaldehyde, 2,3-dihydro-1,4-benzdioxin-5-carbaldehyde |
PubChem CID | 2795033 |
IUPAC Name | 2,3-dihydro-1,4-benzodioxine-5-carbaldehyde |
SMILES | O=CC1=C2OCCOC2=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10679944
|
Thermo Scientific Maybridge
CC01404CB |
250 mg | Amber Glass Bottle |
£54.15
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10356922
|
Thermo Scientific Maybridge
CC01404DA |
1 g | Amber Glass Bottle |
£184.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10452624
|
Thermo Scientific Maybridge
CC01404DE |
5 g | Amber Glass Bottle |
£443.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
29668-43-7 | |
164.16 | |
BJXUCBAQZJITKD-UHFFFAOYSA-N | |
2795033 | |
O=CC1=C2OCCOC2=CC=C1 |
C9H8O3 | |
MFCD00239451 | |
2,3-dihydrobenzo b 1,4 dioxine-5-carbaldehyde, 2,3-dihydro-benzo 1,4 dioxine-5-carbaldehyde, 1,4-benzodioxin-5-carboxaldehyde, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxine-5-carbaldehyd, 2,3-dihydro-1,4-benzodioxin-5-carboxaldehyde, 2,3-ethylenedioxybenzaldehyde, 2,3-dihydrobenzodioxin-5-carboxaldehyde, 1,4-benzodioxane-5-carboxaldehyde, 2h,3h-benzo e 1,4-dioxin-5-carbaldehyde, 2,3-dihydro-1,4-benzdioxin-5-carbaldehyde | |
2,3-dihydro-1,4-benzodioxine-5-carbaldehyde |
Specifications
29668-43-7 | |
C9H8O3 | |
2,3-dihydrobenzo b 1,4 dioxine-5-carbaldehyde, 2,3-dihydro-benzo 1,4 dioxine-5-carbaldehyde, 1,4-benzodioxin-5-carboxaldehyde, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxine-5-carbaldehyd, 2,3-dihydro-1,4-benzodioxin-5-carboxaldehyde, 2,3-ethylenedioxybenzaldehyde, 2,3-dihydrobenzodioxin-5-carboxaldehyde, 1,4-benzodioxane-5-carboxaldehyde, 2h,3h-benzo e 1,4-dioxin-5-carbaldehyde, 2,3-dihydro-1,4-benzdioxin-5-carbaldehyde | |
O=CC1=C2OCCOC2=CC=C1 | |
164.16 | |
164.16 | |
2, 3-Dihydro-1, 4-benzodioxine-5-carbaldehyde |
97% | |
MFCD00239451 | |
BJXUCBAQZJITKD-UHFFFAOYSA-N | |
2,3-dihydro-1,4-benzodioxine-5-carbaldehyde | |
2795033 | |
97% |