missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxine-6-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10099161
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 63758-12-3
Molecular Formula C8H7ClO4S
Molecular Weight (g/mol) 234.65
MDL Number MFCD02677681
InChI Key JWHSRWUHRLYAPM-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzo b 1,4 dioxine-6-sulfonyl chloride, 2,3-dihydro-benzo 1,4 dioxine-6-sulfonyl chloride, 2,3-dihydro-1,4-benzodioxine-6-sulphonylchloride, 2,3-dihydro-benzo 1,4 dioxine-6-sulfonylchloride, 6-chlorosulfonyl-benzo-1,4-dioxane, 1,4-benzodioxin-6-sulfonyl chloride, 2,3-dihydro, 1,4-benzodioxan-6-sulfonyl chloride, 1,4-benzodioxin-6-sulfonylchloride, 2,3-dihydro, 2h,3h-benzo 3,4-e 1,4-dioxin-6-ylchlorosulfone, 6-chlorosulphonyl-2,3-dihydro-1,4-benzodioxine
PubChem CID 2776169
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-sulfonyl chloride
SMILES C1COC2=C(O1)C=CC(=C2)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 234.66
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-6-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.