missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-5-ylboronic acid, 95%, May contain varying amounts of anhydride, Thermo Scientific™

Catalog Number 10190772
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
227305-69-3
C8H9BO3
163.97
MFCD02681979
ZIXLJHSFAMVHPC-UHFFFAOYSA-N
2,3-dihydrobenzofuran-5-boronic acid, 2,3-dihydrobenzofuran-5-ylboronic acid, 2,3-dihydrobenzofuran-5-yl boronic acid, 2,3-dihydrobenzo b furan-5-boronic acid, 2,3-dihydrobenzofuran-5-yl-5-boronic acid, boronic acid, 2,3-dihydro-5-benzofuranyl, 2,3-dihydro-1-benzofuran-5-yl-boranediol, 2,3-dihydro-1-benzofuran-5-yl boronic acid, pubchem7848, zlchem 1108
2773380
OB(O)C1=CC=C2OCCC2=C1
This item is not returnable. View return policy
227305-69-3
C8H9BO3
163.97
MFCD02681979
ZIXLJHSFAMVHPC-UHFFFAOYSA-N
2,3-dihydrobenzofuran-5-boronic acid, 2,3-dihydrobenzofuran-5-ylboronic acid, 2,3-dihydrobenzofuran-5-yl boronic acid, 2,3-dihydrobenzo b furan-5-boronic acid, 2,3-dihydrobenzofuran-5-yl-5-boronic acid, boronic acid, 2,3-dihydro-5-benzofuranyl, 2,3-dihydro-1-benzofuran-5-yl-boranediol, 2,3-dihydro-1-benzofuran-5-yl boronic acid, pubchem7848, zlchem 1108
2773380
OB(O)C1=CC=C2OCCC2=C1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 2,3-dihydro-1-benzofuran-5-ylboronic acid
Formula Weight 163.97
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-5-ylboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'