missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydrobenzo[b]furan-5-ylmethylamine hydrochloride, 97%, Thermo Scientific™
£24.20 - £49.55
Chemical Identifiers
CAS | 55745-74-9 |
---|---|
Molecular Formula | C9H11NO |
Molecular Weight (g/mol) | 149.19 |
MDL Number | MFCD00728869 |
InChI Key | WQXWNTPLZFVZNX-UHFFFAOYSA-N |
Synonym | 5-aminomethyl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-5-yl methanamine, 2,3-dihydrobenzo b furan-5-ylmethylamine, 1-2,3-dihydro-1-benzofuran-5-yl methanamine, 2,3-dihydro-benzofuran-5-ylmethylamine, 2,3-dihydro-1-benzofuran-5-yl methanamine, pubchem6985, 2,3-dihydrobenzofuran-5-ylmethanamine, c-2,3-dihydrobenzofuran-5-ylmethylamine |
PubChem CID | 2735367 |
IUPAC Name | 2,3-dihydro-1-benzofuran-5-ylmethanamine |
SMILES | NCC1=CC2=C(OCC2)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10672053
|
Thermo Scientific Maybridge
CC00813CB |
250 mg | Amber Glass Bottle |
£24.20
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10100782
|
Thermo Scientific Maybridge
CC00813DA |
1 g | Amber Glass Bottle |
£49.55
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
55745-74-9 | |
149.19 | |
WQXWNTPLZFVZNX-UHFFFAOYSA-N | |
2735367 | |
NCC1=CC2=C(OCC2)C=C1 |
C9H11NO | |
MFCD00728869 | |
5-aminomethyl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-5-yl methanamine, 2,3-dihydrobenzo b furan-5-ylmethylamine, 1-2,3-dihydro-1-benzofuran-5-yl methanamine, 2,3-dihydro-benzofuran-5-ylmethylamine, 2,3-dihydro-1-benzofuran-5-yl methanamine, pubchem6985, 2,3-dihydrobenzofuran-5-ylmethanamine, c-2,3-dihydrobenzofuran-5-ylmethylamine | |
2,3-dihydro-1-benzofuran-5-ylmethanamine |
Specifications
55745-74-9 | |
100.0 | |
C9H11NO | |
5-aminomethyl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-5-yl methanamine, 2,3-dihydrobenzo b furan-5-ylmethylamine, 1-2,3-dihydro-1-benzofuran-5-yl methanamine, 2,3-dihydro-benzofuran-5-ylmethylamine, 2,3-dihydro-1-benzofuran-5-yl methanamine, pubchem6985, 2,3-dihydrobenzofuran-5-ylmethanamine, c-2,3-dihydrobenzofuran-5-ylmethylamine | |
NCC1=CC2=C(OCC2)C=C1 | |
149.19 | |
185.65 | |
2, 3-Dihydrobenzo[b]furan-5-ylmethylamine hydrochloride |
96.5 | |
97% | |
MFCD00728869 | |
WQXWNTPLZFVZNX-UHFFFAOYSA-N | |
2,3-dihydro-1-benzofuran-5-ylmethanamine | |
2735367 | |
97% |