missing translation for 'onlineSavingsMsg'
Learn More
2,3-Dihydrobenzo[b]furan-7-ylamine, 97%, Thermo Scientific™ Product Code.: 10404665

2,3-Dihydrobenzo[b]furan-7-ylamine, 97%, Thermo Scientific™

Product Code. 10404665
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10404665

Brand: Thermo Scientific Maybridge CC00914CB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13414-56-7
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD11109313
InChI Key UHHZGSLXPQGPJL-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-7-amine, 2,3-dihydrobenzo b furan-7-ylamine, 2,3-dihydro-benzofuran-7-ylamine, 7-benzofuranamine, 2,3-dihydro, 7-amino-2,3-dihydrobenzofuran, 2,3-dihydro-7-benzofuranamine
PubChem CID 12805950
IUPAC Name 2,3-dihydro-1-benzofuran-7-amine
SMILES NC1=CC=CC2=C1OCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-7-ylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Dihydrobenzo[b]furan-7-ylamine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.