missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydrofuran, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
2,3-Dihydrofuran is a versatile reagent used in lanthanide-catalyzed Diels-Alder reactions with 2-pyrones and in Rh(II)-stabilized cycloadditions with vinylcarbenoids. It is applied for alkylation by active methylene compounds, catalyzed by trans-Dichlorobis(triphenylphosphine)palladium(II). It is also used in the preparation of biologically active compounds such as antitumor agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Dihydrofuran is a versatile reagent used in lanthanide-catalyzed Diels-Alder reactions with 2-pyrones and in Rh(II)-stabilized cycloadditions with vinylcarbenoids. It is applied for alkylation by active methylene compounds, catalyzed by trans-Dichlorobis(triphenylphosphine)palladium(II). It is also used in the preparation of biologically active compounds such as antitumor agents.
Solubility
Soluble in water at 20°C, 20g/L and chloroform.
Notes
Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
2,3-Dihydrofuran is a versatile reagent used in lanthanide-catalyzed Diels-Alder reactions with 2-pyrones and in Rh(II)-stabilized cycloadditions with vinylcarbenoids. It is applied for alkylation by active methylene compounds, catalyzed by trans-Dichlorobis(triphenylphosphine)palladium(II). It is also used in the preparation of biologically active compounds such as antitumor agents.
Solubility
Soluble in water at 20°C, 20g/L and chloroform.
Notes
Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.927 |
Boiling Point | 54°C to 55°C |
Flash Point | −24°C (−11°F) |
Odor | Pungent |
Refractive Index | 1.4245 |
Quantity | 25 g |
UN Number | UN3271 |
Beilstein | 103168 |
Synonym | 4,5-dihydrofuran, furan, 2,3-dihydro, furan, dihydro, dihydrofuran, 2,3 dihydrofuran, dihydrofurane, 2,3dihydrofuran, furan,3-dihydro, 2,3-dihydrofurane, 2,3-dihydro-furan |
Solubility Information | Soluble in water at 20°C,20g/L and chloroform. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'