missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carbonitrile, ≥95%, Thermo Scientific™

Product Code. 10259762
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 859851-02-8
Molecular Formula C7H5NO2S
Molecular Weight (g/mol) 167.18
MDL Number MFCD08060540
InChI Key HSQAPDOVCIFQCU-UHFFFAOYSA-N
Synonym 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbonitrile, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carbonitrile, thieno 3,4-b-1,4-dioxin-5-carbonitrile,2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbonitrile
PubChem CID 7537659
IUPAC Name 2H,3H-thieno[3,4-b][1,4]dioxine-5-carbonitrile
SMILES N#CC1=C2OCCOC2=CS1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 167.19
Percent Purity 95%
Chemical Name or Material 2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'