missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11491127
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40972-86-9
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.98
MDL Number MFCD02683112
InChI Key VREWSCMOGIXMDQ-UHFFFAOYSA-N
Synonym 2,3-dimethoxybenzeneboronic acid, 2,3-dimethoxyphenyl boronic acid, boronic acid, 2,3-dimethoxyphenyl, 2,3-dimethoxyphenylboronicacid, 3-boronoveratrole, pubchem14109, acmc-209jhv, ksc235o1n, 2,3-dimethoxy-phenylboronic acid
PubChem CID 5156491
IUPAC Name (2,3-dimethoxyphenyl)boronic acid
SMILES COC1=CC=CC(B(O)O)=C1OC

Specifications

Melting Point 67°C to 71°C
Quantity 1 g
Beilstein 3051295
Formula Weight 181.98
Percent Purity 98%
Chemical Name or Material 2,3-Dimethoxybenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'