missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzonitrile, 98%, Thermo Scientific™

Product Code. 11469996
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed in the replacement of nuclear alkoxyl groups to yield dimethoxy ketones by the action of methyl and phenyl Grignard reagents on 2,3-dimethoxybenzonitrile.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5653-62-3
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00001784
InChI Key LBXGBNHUNHWYRM-UHFFFAOYSA-N
Synonym o-veratronitrile, benzonitrile, 2,3-dimethoxy, 2,3-dimethoxybenzenecarbonitrile, unii-g1te317ghw, 2,3-dimethoxy benzonitrile, g1te317ghw, pubchem13104, o-veratronitrile 8ci, benzonitrile,3-dimethoxy, acmc-1aul4
PubChem CID 21862
IUPAC Name 2,3-dimethoxybenzonitrile
SMILES COC1=CC=CC(=C1OC)C#N

Specifications

Melting Point 44°C to 47°C
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN3439
Beilstein 2719631
Solubility Information Insoluble in water.
Formula Weight 163.18
Percent Purity 98%
Chemical Name or Material 2,3-Dimethoxybenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'