Learn More
2-[3-(Dimethylamino)propoxy]aniline, 97%, Thermo Scientific™
£78.20 - £187.00
Chemical Identifiers
CAS | 1134-76-5 |
---|---|
Molecular Formula | C11H18N2O |
Molecular Weight (g/mol) | 194.278 |
MDL Number | MFCD08687610 |
InChI Key | XGTIDBFMHOJXSS-UHFFFAOYSA-N |
Synonym | 2-3-dimethylamino propoxy aniline, benzenamine, 2-3-dimethylamino propoxy, 2-3-dimethylaminopropoxy aniline, 3-2-aminophenoxy propyl dimethylamine, 2-3-dimethylamino-propoxy-phenylamineShow More |
PubChem CID | 18672712 |
IUPAC Name | 2-[3-(dimethylamino)propoxy]aniline |
SMILES | CN(C)CCCOC1=CC=CC=C1N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10193174
|
Thermo Scientific Maybridge
CC59614CB |
250 mg | Amber Glass Bottle |
£78.20
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10022114
|
Thermo Scientific Maybridge
CC59614DA |
1 g | Amber Glass Bottle |
£187.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1134-76-5 | |
194.278 | |
XGTIDBFMHOJXSS-UHFFFAOYSA-N | |
18672712 | |
CN(C)CCCOC1=CC=CC=C1N |
C11H18N2O | |
MFCD08687610 | |
2-3-dimethylamino propoxy aniline, benzenamine, 2-3-dimethylamino propoxy, 2-3-dimethylaminopropoxy aniline, 3-2-aminophenoxy propyl dimethylamine, 2-3-dimethylamino-propoxy-phenylamineShow More | |
2-[3-(dimethylamino)propoxy]aniline |
Specifications
1134-76-5 | |
100.0 | |
C11H18N2O | |
2-3-dimethylamino propoxy aniline, benzenamine, 2-3-dimethylamino propoxy, 2-3-dimethylaminopropoxy aniline, 3-2-aminophenoxy propyl dimethylamine, 2-3-dimethylamino-propoxy-phenylamine | |
CN(C)CCCOC1=CC=CC=C1N | |
194.278 | |
194.28 | |
2-[3-(Dimethylamino)propoxy]aniline |
96.5 | |
97% | |
MFCD08687610 | |
XGTIDBFMHOJXSS-UHFFFAOYSA-N | |
2-[3-(dimethylamino)propoxy]aniline | |
18672712 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.