missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Dimethylamino)propoxy]benzonitrile, 97%, Thermo Scientific™

Product Code. 10103184
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 910037-05-7
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD09064997
InChI Key IRLARMCZZWFRCP-UHFFFAOYSA-N
Synonym 2-3-dimethylamino propoxy benzonitrile, 2-3-dimethylamino propoxy benzenecarbonitrile
PubChem CID 2993332
IUPAC Name 2-[3-(dimethylamino)propoxy]benzonitrile
SMILES CN(C)CCCOC1=CC=CC=C1C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 2-[3-(Dimethylamino)propoxy]benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'