missing translation for 'onlineSavingsMsg'
Learn More

(2,3-Dimethylphenoxy)acetic acid, 98%, Thermo Scientific™

Product Code. 10484833
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2935-63-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00016814
InChI Key AVDBMBYECMTQJQ-UHFFFAOYSA-N
Synonym 2,3-dimethylphenoxyacetic acid, 2-2,3-dimethylphenoxy acetic acid, 2,3-dimethylphenoxy acetic acid, 2,3-xylyloxyacetic acid, acmc-20aog1, enamine_005510, 2,3-dimethylphenoxyessig, cambridge id 5161587, 2,3-dimethylphenoxy acetic acid, acetic acid, 2,3-dimethylphenoxy
PubChem CID 76239
IUPAC Name 2-(2,3-dimethylphenoxy)acetic acid
SMILES CC1=CC=CC(OCC(O)=O)=C1C

Specifications

Melting Point 186°C to 190°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2C6H3OCH2CO2H
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material (2, 3-Dimethylphenoxy)acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'