missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylpiperazine, cis + trans, 95%, Thermo Scientific Chemicals

Product Code. 11346719
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84468-52-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD07373397
InChI Key COWPTMLRSANSMQ-UHFFFAOYSA-N
Synonym 2,3-dimethyl-piperazine, piperazine, 2,3-dimethyl, 2,3-dimethylpiperazine, 2,3-dimethylpiperazine, cis + trans
PubChem CID 522814
IUPAC Name 2,3-dimethylpiperazine
SMILES CC1C(NCCN1)C

Specifications

Melting Point 81°C to 90°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 114.19
Percent Purity 95%
Chemical Name or Material 2,3-Dimethylpiperazine, cis + trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'