missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water, Thermo Scientific Chemicals
£35.20 - £437.00
Chemical Identifiers
CAS | 886-38-4 |
---|---|
Molecular Formula | C15H10O |
Molecular Weight (g/mol) | 206.24 |
MDL Number | MFCD00001311 |
InChI Key | HCIBTBXNLVOFER-UHFFFAOYSA-N |
Synonym | diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec |
PubChem CID | 65057 |
ChEBI | CHEBI:53074 |
IUPAC Name | 2,3-diphenylcycloprop-2-en-1-one |
SMILES | O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11475057
|
Thermo Scientific Alfa Aesar
B21954.03 |
1 g |
£35.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485057
|
Thermo Scientific Alfa Aesar
B21954.06 |
5 g |
£113.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11495057
|
Thermo Scientific Alfa Aesar
B21954.14 |
25 g |
£437.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
886-38-4 | |
206.24 | |
HCIBTBXNLVOFER-UHFFFAOYSA-N | |
65057 | |
2,3-diphenylcycloprop-2-en-1-one |
C15H10O | |
MFCD00001311 | |
diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec | |
CHEBI:53074 | |
O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
886-38-4 | |
C15H10O | |
608049 | |
diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec | |
O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1 | |
206.24 | |
CHEBI:53074 | |
99% | |
2,3-Diphenyl-2-cyclopropen-1-one |
119°C to 121°C | |
MFCD00001311 | |
14,3308 | |
HCIBTBXNLVOFER-UHFFFAOYSA-N | |
2,3-diphenylcycloprop-2-en-1-one | |
65057 | |
206.24 | |
(dry wt.), may cont up to 5% water |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
missing translation for 'einecsNumber' : 212-948-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only