missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Fluorophenyl)ethylamine, ≥97%, Thermo Scientific Chemicals

Artikelnummer. 15402109
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 404-70-6
Molecular Formula C8H11FN
Molecular Weight (g/mol) 140.18
MDL Number MFCD00075376
InChI Key AUCVZEYHEFAWHO-UHFFFAOYSA-O
Synonym 3-fluorophenethylamine, 2-3-fluorophenyl ethylamine, 2-3-fluoro-phenyl-ethylamine, 2-3-fluorophenyl ethanamine, 2-3-fluorophenyl ethan-1-amine, benzeneethanamine, 3-fluoro, m-fluorophenethylamine, 3-flourophenethylamine, m-fluorophenylethylamine, 3-fluoro phenethylamine
PubChem CID 533928
IUPAC Name 2-(3-fluorophenyl)ethanamine
SMILES [NH3+]CCC1=CC=CC(F)=C1

Spezifikation

Density 1.066 g/mL
Boiling Point 87°C (15 mmHg)
Flash Point 83°C (181°F)
Refractive Index 1.509
Quantity 100 g
UN Number UN2735
Beilstein 3045789
Formula Weight 139.17
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 2-(3-Fluorophenyl)ethylamine
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'