missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Hexanedione, 94%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.
Solubility
Partly miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.
Solubility
Partly miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -30°C |
Density | 0.937 |
Boiling Point | 127°C to 128°C |
Flash Point | 28°C (82°F) |
Odor | Characteristic |
Refractive Index | 1.412 |
Quantity | 5 g |
UN Number | UN1224 |
Beilstein | 1699896 |
Solubility Information | Partly miscible in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'