Learn More
2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals
POA - £281.40
Chemical Identifiers
CAS | 2039-67-0 |
---|---|
Molecular Formula | C9H13NO |
Molecular Weight (g/mol) | 151.209 |
MDL Number | MFCD00008187 |
InChI Key | WJBMRZAHTUFBGE-UHFFFAOYSA-N |
Synonym | 3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine |
PubChem CID | 74866 |
IUPAC Name | 2-(3-methoxyphenyl)ethanamine |
SMILES | COC1=CC=CC(=C1)CCN |
Description
Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsActivated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
2039-67-0 | |
151.209 | |
WJBMRZAHTUFBGE-UHFFFAOYSA-N | |
74866 | |
COC1=CC=CC(=C1)CCN |
C9H13NO | |
MFCD00008187 | |
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine | |
2-(3-methoxyphenyl)ethanamine |
Specifications
2039-67-0 | |
1.038 g/mL | |
>110°C (230°F) | |
1.538 | |
UN2735 | |
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine | |
WJBMRZAHTUFBGE-UHFFFAOYSA-N | |
2-(3-methoxyphenyl)ethanamine | |
74866 | |
≥97% | |
2-(3-Methoxyphenyl)ethylamine |
Yellow | |
118°C to 118°C (6 mmHg) | |
C9H13NO | |
MFCD00008187 | |
775202 | |
Insoluble in water. | |
COC1=CC=CC(=C1)CCN | |
151.209 | |
151.21 | |
Liquid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 218-017-9
missing translation for 'rtecsNumber' : SH7835000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.