missing translation for 'onlineSavingsMsg'
Learn More

2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole, 98%, Thermo Scientific Chemicals

Product Code. 15445535
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103577-40-8
Molecular Formula C16H14F3N3OS
Molecular Weight (g/mol) 353.363
MDL Number MFCD00834357
InChI Key CCHLMSUZHFPSFC-UHFFFAOYSA-N
Synonym lansoprazole sulfide, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridin-2-yl methyl thio-1h-benzo d imidazole, 2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methylthio-1h-benzimidazole, unii-58xp950e2t, lansoprazole sulphide compound, 2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methyl thio-1h-benzimidazole, 2-methyl-4-2,2,2-trifluoroethoxy pyridinyl thio-1h-binzimidazole, 1h-benzimidazole,2-3-methyl-4-2,2,2-trifluoroethoxy-2-pyridinyl methyl thio, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridin-2-yl methyl sulfanyl-1h-1,3-benzodiazole, 2-3-methyl-4-2,2,2-trifluoroethoxy pyridinyl methyl thio-1h-benzimidazole
PubChem CID 1094080
IUPAC Name 2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfanyl]-1H-benzimidazole
SMILES CC1=C(C=CN=C1CSC2=NC3=CC=CC=C3N2)OCC(F)(F)F

Specifications

Quantity 5 g
Formula Weight 353.36
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'