missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Methylphenyl)thioacetamide, 97%, Thermo Scientific™

Product Code. 15415817
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6487-91-8
Molecular Formula C9H11NS
Molecular Weight (g/mol) 165.25
MDL Number MFCD09757580
InChI Key APDOZLDJDPJFLM-UHFFFAOYSA-N
Synonym 2-3-methylphenyl ethanethioamide, 2-m-tolyl thioacetamide, 2-m-tolyl ethanethioamide, 3-methylphenyl thioacetamide, 3-methylphenyl ethanethioamide, benzeneethanethioamide,3-methyl, 2-3-methylphenyl thioacetamide, benzeneethanethioamide, 3-methyl
PubChem CID 21504242
IUPAC Name 2-(3-methylphenyl)ethanethioamide
SMILES CC1=CC(CC(N)=S)=CC=C1

Specifications

Melting Point 120°C to 124°C
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 165.26
Percent Purity 97%
Chemical Name or Material 2-(3-Methylphenyl)thioacetamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'