missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2',3'-O-Isopropylideneadenosine, 98%
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Absorbance | 0.75 to 0.81 (A250/A260),0.13 to 0.17 (A280/A260),0.01 (A290/A260) max |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 221.0°C to 222.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Quantity | 25 g |
Spectrophotometry | 96% min. (lambda max. 259nm,pH 7) molar absorptivity 15400+/-4%,on dry substance |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'