missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pentanedione, 97%, Thermo Scientific Chemicals

Product Code. 10553771
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-14-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009313
InChI Key TZMFJUDUGYTVRY-UHFFFAOYSA-N
Synonym 2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone
PubChem CID 11747
ChEBI CHEBI:52774
IUPAC Name pentane-2,3-dione
SMILES CCC(=O)C(C)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -52.0°C
Color Yellow
Density 0.9580g/mL
Boiling Point 110.0°C to 112.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5COCOCH3
Refractive Index 1.4020 to 1.4060
Quantity 100 mL
Beilstein 01,776
Specific Gravity 0.958
Solubility Information Solubility in water: 60g/L (15°C). Other solubilities: miscible with ethanol, glycerol and, propylene glycol, ether
Formula Weight 100.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3-Pentanedione, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Pentanedione
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.