missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pentanedione, 97%, Thermo Scientific Chemicals

Product Code. 11404464
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-14-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009313
InChI Key TZMFJUDUGYTVRY-UHFFFAOYSA-N
Synonym 2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone
PubChem CID 11747
ChEBI CHEBI:52774
IUPAC Name pentane-2,3-dione
SMILES CCC(=O)C(C)=O

Specifications

Melting Point -52°C
Boiling Point 110°C to 112°C
Flash Point 18°C (64°F)
Odor Odorless
Quantity 100 g
UN Number UN1224
Beilstein 1699638
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material 2,3-Pentanedione
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'