missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Pentanedione, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -52°C |
Boiling Point | 110°C to 112°C |
Flash Point | 18°C (64°F) |
Odor | Odorless |
Quantity | 25 g |
UN Number | UN1224 |
Beilstein | 1699638 |
Formula Weight | 100.12 |
Percent Purity | 97% |
Chemical Name or Material | 2,3-Pentanedione |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 2
- H225-Highly flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'