Learn More
2,3-Pentanedione, 97%, Thermo Scientific Chemicals
£30.60 - £294.00
Chemical Identifiers
CAS | 600-14-6 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00009313 |
InChI Key | TZMFJUDUGYTVRY-UHFFFAOYSA-N |
Synonym | 2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone |
PubChem CID | 11747 |
ChEBI | CHEBI:52774 |
IUPAC Name | pentane-2,3-dione |
SMILES | CCC(=O)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494454
|
Thermo Scientific Alfa Aesar
A13116.14 |
25 g |
£30.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404464
|
Thermo Scientific Alfa Aesar
A13116.22 |
100 g |
£78.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414464
|
Thermo Scientific Alfa Aesar
A13116.36 |
500 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
600-14-6 | |
100.12 | |
TZMFJUDUGYTVRY-UHFFFAOYSA-N | |
11747 | |
pentane-2,3-dione |
C5H8O2 | |
MFCD00009313 | |
2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone | |
CHEBI:52774 | |
CCC(=O)C(C)=O |
Specifications
600-14-6 | |
110°C to 112°C | |
Odorless | |
MFCD00009313 | |
1699638 | |
TZMFJUDUGYTVRY-UHFFFAOYSA-N | |
pentane-2,3-dione | |
11747 | |
100.12 | |
2,3-Pentanedione |
-52°C | |
18°C (64°F) | |
C5H8O2 | |
UN1224 | |
2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone | |
CCC(=O)C(C)=O | |
100.12 | |
CHEBI:52774 | |
97% |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
missing translation for 'rtecsNumber' : SA1850000
missing translation for 'tsca' : TSCA
Recommended Storage : 2° to 8°C
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only