missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Phenylpropyl)pyridine, 98%, Thermo Scientific™

Product Code. 10749023
Click to view available options
1g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 2110-18-1
CAS Min % 97.5
CAS Max % 100.0
Density 1.0150g/mL
Boiling Point 115.0°C (2.0 mmHg)
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C14H15N
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
MDL Number MFCD00191237
Quantity 1g
Specific Gravity 1.015
Synonym 2-3-phenylpropyl pyridine, pyridine, 2-3-phenylpropyl, unii-9f0bfm744f, alpha-3-phenylpropyl pyridine, 2-phenylpropylpyridine, cortex pyridine, phenylpropylpyridine, 3-phenylpropyl pyridine, acmc-1cs6o, dsstox_cid_22356
InChI Key JJJPNTQYUJPWGQ-UHFFFAOYSA-N
SMILES C(CC1=CC=CC=C1)CC1=CC=CC=N1
IUPAC Name 2-(3-phenylpropyl)pyridine
Molecular Weight (g/mol) 197.28
PubChem CID 459494
Formula Weight 197.28
Percent Purity 98%
Chemical Name or Material 2-(3-Phenylpropyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-(3-Phenylpropyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'