missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,4-Trimethyl-1-pentene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
2,4,4-Trimethyl-1-pentene is used in the manufacturing of chemicals, adhesives, sealant chemicals, fuels and fuel additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,4-Trimethyl-1-pentene is used in the manufacturing of chemicals, adhesives, sealant chemicals, fuels and fuel additives.
Solubility
Miscible with acetone. Partially soluble in methanol, diethyl ether, benzene, chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agent.
2,4,4-Trimethyl-1-pentene is used in the manufacturing of chemicals, adhesives, sealant chemicals, fuels and fuel additives.
Solubility
Miscible with acetone. Partially soluble in methanol, diethyl ether, benzene, chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agent.
Specifications
Melting Point | -94°C |
Density | 0.715 |
Boiling Point | 101°C to 102°C |
Flash Point | −6°C (21°F) |
Odor | Petrol-like |
Refractive Index | 1.4085 |
Quantity | 25 g |
UN Number | UN2050 |
Beilstein | 1098309 |
Solubility Information | Miscible with acetone. Partially soluble in methanol,diethyl ether,benzene,chloroform. Immiscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'