Learn More
2,4,5-Tribromoimidazole, 97%, Thermo Scientific™
£22.23
Chemical Identifiers
CAS | 2034-22-2 |
---|---|
MDL Number | MFCD00005184 |
InChI Key | JCGGPCDDFXIVQB-UHFFFAOYSA-N |
Synonym | 2,4,5-tribromoimidazole, 1h-imidazole, 2,4,5-tribromo, imidazole, 2,4,5-tribromo, 2,4,5-tribromoimidazo, tribromoimidazole, pubchem7150, 2,5-tribromoimidazole, acmc-1cmbx, timtec-bb sbb001252, imidazole,4,5-tribromo |
PubChem CID | 16253 |
IUPAC Name | 2,4,5-tribromo-1H-imidazole |
SMILES | C1(=C(N=C(N1)Br)Br)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10173790
|
Thermo Scientific Acros
161640050 |
5 g | Glass bottle | ||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2034-22-2 | |
JCGGPCDDFXIVQB-UHFFFAOYSA-N | |
16253 | |
C1(=C(N=C(N1)Br)Br)Br |
MFCD00005184 | |
2,4,5-tribromoimidazole, 1h-imidazole, 2,4,5-tribromo, imidazole, 2,4,5-tribromo, 2,4,5-tribromoimidazo, tribromoimidazole, pubchem7150, 2,5-tribromoimidazole, acmc-1cmbx, timtec-bb sbb001252, imidazole,4,5-tribromo | |
2,4,5-tribromo-1H-imidazole |
Specifications
2034-22-2 | |
100.0 | |
Beige-Yellow | |
97% | |
MFCD00005184 | |
2,4,5-tribromoimidazole, 1h-imidazole, 2,4,5-tribromo, imidazole, 2,4,5-tribromo, 2,4,5-tribromoimidazo, tribromoimidazole, pubchem7150, 2,5-tribromoimidazole, acmc-1cmbx, timtec-bb sbb001252, imidazole,4,5-tribromo | |
C1(=C(N=C(N1)Br)Br)Br | |
304.76 | |
304.76 | |
Powder |
96.0 | |
208.0°C to 212.0°C | |
Authentic | |
C3HBr3N2 | |
23, 49 | |
JCGGPCDDFXIVQB-UHFFFAOYSA-N | |
2,4,5-tribromo-1H-imidazole | |
16253 | |
97% | |
2, 4, 5-Tribromoimidazole |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 217-997-5
- 2,4,5-Tribromoimidazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /