Learn More
2,4,5-Trimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals
£39.10 - £108.00
Chemical Identifiers
CAS | 4460-86-0 |
---|---|
Molecular Formula | C10H12O4 |
Molecular Weight (g/mol) | 196.20 |
MDL Number | MFCD00003312 |
InChI Key | IAJBQAYHSQIQRE-UHFFFAOYSA-N |
Synonym | asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296 |
PubChem CID | 20525 |
IUPAC Name | 2,4,5-trimethoxybenzaldehyde |
SMILES | COC1=CC(OC)=C(C=O)C=C1OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10173300
|
Thermo Scientific Acros
157940250 |
25 g | Glass bottle |
£39.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10469260
|
Thermo Scientific Acros
157941000 |
100 g | Glass bottle |
£108.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4460-86-0 | |
196.20 | |
IAJBQAYHSQIQRE-UHFFFAOYSA-N | |
20525 | |
COC1=CC(OC)=C(C=O)C=C1OC |
C10H12O4 | |
MFCD00003312 | |
asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296 | |
2,4,5-trimethoxybenzaldehyde |
Specifications
4460-86-0 | |
100.0 | |
White to Beige | |
Authentic | |
C10H12O4 | |
MFCD00003312 | |
asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296 | |
COC1=CC(OC)=C(C=O)C=C1OC | |
196.20 | |
196.2 | |
Fine Crystalline Powder |
97.5 | |
111.0°C to 115.0°C | |
140°C (4.0 mmHg) | |
98% | |
(CH3O)3C6H2CHO | |
08, 389 | |
IAJBQAYHSQIQRE-UHFFFAOYSA-N | |
2,4,5-trimethoxybenzaldehyde | |
20525 | |
98% | |
2, 4, 5-Trimethoxybenzaldehyde, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 224-713-3
missing translation for 'rtecsNumber' : CU8460000
missing translation for 'tsca' : TSCA
- 2,4,5-Trimethoxybenzaldehyde
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only