missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethyloxazole, 97%, Thermo Scientific Chemicals

Product Code. 11484093
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20662-84-4
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.144
MDL Number MFCD00005308
InChI Key ZRLDBDZSLLGDOX-UHFFFAOYSA-N
Synonym 2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636
PubChem CID 30215
IUPAC Name 2,4,5-trimethyl-1,3-oxazole
SMILES CC1=C(OC(=N1)C)C

Specifications

Density 0.964
Boiling Point 133°C to 134°C
Flash Point 33°C (91°F)
Odor Pungent
Refractive Index 1.442
Quantity 25 g
UN Number UN1993
Beilstein 107043
Formula Weight 111.14
Percent Purity 97%
Chemical Name or Material 2,4,5-Trimethyloxazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'