missing translation for 'onlineSavingsMsg'
Learn More

2,4,6,8-Tetramethylcyclotetrasiloxane, 99%, Thermo Scientific Chemicals

Catalog Number 11359295
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2370-88-9
C4H12O4Si4
236.476
MFCD00039567
WZJUBBHODHNQPW-UHFFFAOYSA-N
2,4,6,8-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethylcyclotetrasiloxane, tetramethylcyclotetrasiloxane, tmcts, cyclotetrasiloxane, 2,4,6,8-tetramethyl, 2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 1,2,5,7-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-cyclotetrasiloxan, 2,4,6,8-tetramethylcyclotetrasiloxne, 1,3,5,7-tetramethyl cyclotetrasiloxane
6327421
2,4,6,8-tetramethyl-5,7-dioxa-1,3-dioxonia-2,4-disila-6,8-disilanidacycloocta-1,3-diene
C[Si-]1O[Si-]([O+]=[Si]([O+]=[Si](O1)C)C)C
This item is not returnable. View return policy
2370-88-9
C4H12O4Si4
236.476
MFCD00039567
WZJUBBHODHNQPW-UHFFFAOYSA-N
2,4,6,8-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethylcyclotetrasiloxane, tetramethylcyclotetrasiloxane, tmcts, cyclotetrasiloxane, 2,4,6,8-tetramethyl, 2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 1,2,5,7-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-cyclotetrasiloxan, 2,4,6,8-tetramethylcyclotetrasiloxne, 1,3,5,7-tetramethyl cyclotetrasiloxane
6327421
2,4,6,8-tetramethyl-5,7-dioxa-1,3-dioxonia-2,4-disila-6,8-disilanidacycloocta-1,3-diene
C[Si-]1O[Si-]([O+]=[Si]([O+]=[Si](O1)C)C)C

Tetramethyl-cyclotetrasiloxane is a good impregnant of photoelectric material. It is used to manufacture of modified siloxane with defined hydrogen content and chain quantity. It is also used in production of silicone polymers and acts as a precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics and semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl-cyclotetrasiloxane is a good impregnant of photoelectric material. It is used to manufacture of modified siloxane with defined hydrogen content and chain quantity. It is also used in production of silicone polymers and acts as a precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics and semiconductors.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents, bases and acids. Store in a cool place.
TRUSTED_SUSTAINABILITY
Boiling Point 134°C to 135°C
Melting Point -69°C
Quantity 5 g
UN Number UN1993
Beilstein 1787074
Flash Point 25°C (77°F)
Formula Weight 240.51
Percent Purity 99%
Chemical Name or Material 2,4,6,8-Tetramethylcyclotetrasiloxane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'