missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Collidine, 98+%, Pure, Fisher Chemical
Click to view available options
100mL, Plastic Coated Glass bottle
Specifications
CAS | 108-75-8 |
Melting Point | -43°C |
Color | Yellow |
Boiling Point | 171°C |
Packaging | Plastic Coated Glass bottle |
Molecular Formula | C8H11N |
MDL Number | 6338 |
Quantity | 100 mL |
Formula Weight | 121.18g/mol |
Vapor Pressure | 4 hPa at 20°C |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
Product Identifier
- 2,4,6-Collidine
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'