Learn More
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -43.0°C |
Color | Colorless to Yellow |
Density | 0.9100g/mL |
Boiling Point | 171.0°C to 172.0°C |
Flash Point | 57°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0463991 | Certificate of Analysis | 21/06/2024 | 161955000,161950025,161951000,161950010 |
A0463131 | Certificate of Analysis | 23/05/2024 | 161955000,161950025,161951000,161950010 |
A0459860 | Certificate of Analysis | 02/02/2024 | 161955000,161950025,161951000,161950010 |
A0446716 | Certificate of Analysis | 13/10/2023 | 161955000,161950025,161951000,161950010 |
A0438811 | Certificate of Analysis | 06/02/2023 | 161955000,161950025,161951000,161950010 |
Safety and Handling


- 2,4,6-Collidine
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.