missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.
Solubility
Soluble in methanol.
Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.
Solubility
Soluble in methanol.
Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
Specifications
Melting Point | 248°C to 252°C |
Quantity | 5 g |
Beilstein | 282581 |
Merck Index | 14,9750 |
Solubility Information | Soluble in methanol. |
IUPAC Name | 2,4,6-tripyridin-2-yl-1,3,5-triazine |
Formula Weight | 312.34 |
Percent Purity | 98% |
Chemical Name or Material | 2,4,6-Tri(2-pyridyl)-1,3,5-triazine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'