Learn More
2,4,6-Tri-tert-butylphenol, 97%, Thermo Scientific Chemicals
£16.70 - £239.00
Chemical Identifiers
CAS | 732-26-3 |
---|---|
Molecular Formula | C18H30O |
Molecular Weight (g/mol) | 262.44 |
MDL Number | MFCD00008821 |
InChI Key | PFEFOYRSMXVNEL-UHFFFAOYSA-N |
Synonym | 2,4,6-tri-tert-butylphenol, voidox, alkofen b, 2,4,6-tris tert-butyl phenol, 2,4,6-tri-t-butylphenol, phenol, 2,4,6-tris 1,1-dimethylethyl, phenol, 2,4,6-tri-tert-butyl, 2,4,6-tris 1,1-dimethylethyl phenol, unii-99j2vic675, tris 1,1-dimethylethyl phenol |
PubChem CID | 12902 |
SMILES | CC(C)(C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10419052
|
Thermo Scientific Acros
139330050 |
5 g | Glass Bottle |
£16.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10525401
|
Thermo Scientific Acros
139330500 |
50 g | Plastic bottle |
£33.30
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10510234
|
Thermo Scientific Acros
139335000 |
500 g | Plastic bottle |
£239.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
732-26-3 | |
262.44 | |
PFEFOYRSMXVNEL-UHFFFAOYSA-N | |
12902 |
C18H30O | |
MFCD00008821 | |
2,4,6-tri-tert-butylphenol, voidox, alkofen b, 2,4,6-tris tert-butyl phenol, 2,4,6-tri-t-butylphenol, phenol, 2,4,6-tris 1,1-dimethylethyl, phenol, 2,4,6-tri-tert-butyl, 2,4,6-tris 1,1-dimethylethyl phenol, unii-99j2vic675, tris 1,1-dimethylethyl phenol | |
CC(C)(C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C |
Specifications
732-26-3 | |
100.0 | |
277°C | |
C18H30O | |
MFCD00008821 | |
06, IV, 3539 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
CC(C)(C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C | |
262.44 | |
262.42 | |
2, 4, 6-Tri-tert-butylphenol |
96.0 | |
129°C to 132°C | |
96% min. (GC) | |
[(CH3)3C]3C6H2OH | |
3077 | |
2,4,6-tri-tert-butylphenol, voidox, alkofen b, 2,4,6-tris tert-butyl phenol, 2,4,6-tri-t-butylphenol, phenol, 2,4,6-tris 1,1-dimethylethyl, phenol, 2,4,6-tri-tert-butyl, 2,4,6-tris 1,1-dimethylethyl phenol, unii-99j2vic675, tris 1,1-dimethylethyl phenol | |
PFEFOYRSMXVNEL-UHFFFAOYSA-N | |
2,4,6-tritert-butylphenol | |
12902 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If s
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 211-989-5
RUO – Research Use Only