Learn More
2,4,6-Tribromophenol, 98%, Thermo Scientific Chemicals
£40.50 - £113.57
Chemical Identifiers
CAS | 118-79-6 |
---|---|
Molecular Formula | C6H3Br3O |
Molecular Weight (g/mol) | 330.80 |
MDL Number | MFCD00002150 |
InChI Key | BSWWXRFVMJHFBN-UHFFFAOYSA-N |
Synonym | tribromophenol, bromol, bromkal pur 3, xeroform, phenol, 2,4,6-tribromo, flammex 3bp, unii-ys6k3eu393, ccris 1658, great lakes ph-73, 5175-83-7 bismuth 3+ salt |
PubChem CID | 1483 |
ChEBI | CHEBI:47696 |
IUPAC Name | 2,4,6-tribromophenol |
SMILES | OC1=C(Br)C=C(Br)C=C1Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439623
|
Thermo Scientific Alfa Aesar
A12424.30 |
250 g |
£40.50
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429623
|
Thermo Scientific Alfa Aesar
A12424.0B |
1000 g |
£113.57
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
118-79-6 | |
330.80 | |
BSWWXRFVMJHFBN-UHFFFAOYSA-N | |
1483 | |
2,4,6-tribromophenol |
C6H3Br3O | |
MFCD00002150 | |
tribromophenol, bromol, bromkal pur 3, xeroform, phenol, 2,4,6-tribromo, flammex 3bp, unii-ys6k3eu393, ccris 1658, great lakes ph-73, 5175-83-7 bismuth 3+ salt | |
CHEBI:47696 | |
OC1=C(Br)C=C(Br)C=C1Br |
Specifications
118-79-6 | |
2.55 | |
C6H3Br3O | |
UN3077 | |
14,9613 | |
BSWWXRFVMJHFBN-UHFFFAOYSA-N | |
2,4,6-tribromophenol | |
1483 | |
330.8 | |
2,4,6-Tribromophenol |
86°C to 92°C | |
282°C to 290°C (sublimation) | |
MFCD00002150 | |
776920 | |
tribromophenol, bromol, bromkal pur 3, xeroform, phenol, 2,4,6-tribromo, flammex 3bp, unii-ys6k3eu393, ccris 1658, great lakes ph-73, 5175-83-7 bismuth 3+ salt | |
OC1=C(Br)C=C(Br)C=C1Br | |
330.80 | |
CHEBI:47696 | |
98% |
Safety and Handling
GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
H317-H319
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 204-278-6
missing translation for 'rtecsNumber' : SN1225000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.