Learn More
2,4,6-Trifluorobenzoyl chloride, 97%, Thermo Scientific Chemicals
£35.70 - £133.00
Chemical Identifiers
CAS | 79538-29-7 |
---|---|
Molecular Formula | C7H2ClF3O |
Molecular Weight (g/mol) | 194.537 |
MDL Number | MFCD00070810 |
InChI Key | SIFIJQFBERMWMU-UHFFFAOYSA-N |
Synonym | 2,4,6-trifluorobenzoylchloride, 2,4,6-trifluoro-benzoyl chloride, acmc-20aof3, timtec-bb sbb005827, 2,4,6-tri-fluorobenzoyl chloride, 2,4,6-trifluorobenzoyl chloride, benzoyl chloride,2,4,6-trifluoro, benzoyl chloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzoyl chloride 1g, benzoyl chloride, 2,4,6-trifluoro-9ci |
PubChem CID | 144895 |
IUPAC Name | 2,4,6-trifluorobenzoyl chloride |
SMILES | C1=C(C=C(C(=C1F)C(=O)Cl)F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449846
|
Thermo Scientific Alfa Aesar
B20323.03 |
1 g |
£35.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459846
|
Thermo Scientific Alfa Aesar
B20323.06 |
5 g |
£133.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79538-29-7 | |
194.537 | |
SIFIJQFBERMWMU-UHFFFAOYSA-N | |
144895 | |
C1=C(C=C(C(=C1F)C(=O)Cl)F)F |
C7H2ClF3O | |
MFCD00070810 | |
2,4,6-trifluorobenzoylchloride, 2,4,6-trifluoro-benzoyl chloride, acmc-20aof3, timtec-bb sbb005827, 2,4,6-tri-fluorobenzoyl chloride, 2,4,6-trifluorobenzoyl chloride, benzoyl chloride,2,4,6-trifluoro, benzoyl chloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzoyl chloride 1g, benzoyl chloride, 2,4,6-trifluoro-9ci | |
2,4,6-trifluorobenzoyl chloride |
Specifications
79538-29-7 | |
167°C to 168°C | |
1.484 | |
UN1760 | |
Moisture sensitive | |
SIFIJQFBERMWMU-UHFFFAOYSA-N | |
2,4,6-trifluorobenzoyl chloride | |
144895 | |
97% |
1.499 | |
C7H2ClF3O | |
MFCD00070810 | |
1958563 | |
2,4,6-trifluorobenzoylchloride, 2,4,6-trifluoro-benzoyl chloride, acmc-20aof3, timtec-bb sbb005827, 2,4,6-tri-fluorobenzoyl chloride, 2,4,6-trifluorobenzoyl chloride, benzoyl chloride,2,4,6-trifluoro, benzoyl chloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzoyl chloride 1g, benzoyl chloride, 2,4,6-trifluoro-9ci | |
C1=C(C=C(C(=C1F)C(=O)Cl)F)F | |
194.537 | |
194.54 | |
2,4,6-Trifluorobenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only