missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide, Thermo Scientific Chemicals

Product Code. 15460437
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

An inactive analog of m-3M3FBS. Used as a negative control.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An inactive analog of m-3M3FBS
  • Used as a negative control
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313981-55-4
Molecular Formula C16H16F3NO2S
Molecular Weight (g/mol) 343.36
MDL Number MFCD00784362
InChI Key SKJJIFRWCCSXGL-UHFFFAOYSA-N
Synonym o-3m3fbs, 2,4,6-trimethyl-n-2-trifluoromethyl phenyl benzenesulfonamide, tocris-1942, benzenesulfonamide,2,4,6-trimethyl-n-2-trifluoromethyl phenyl
PubChem CID 770820
IUPAC Name 2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
SMILES CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F

Specifications

Color White
Quantity 50 mg
Formula Weight 343.36
Physical Form Powder
Chemical Name or Material 2,4,6-Trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'