missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide, Thermo Scientific Chemicals
£83.10 - £306.00
Chemical Identifiers
CAS | 313981-55-4 |
---|---|
Molecular Formula | C16H16F3NO2S |
Molecular Weight (g/mol) | 343.36 |
MDL Number | MFCD00784362 |
InChI Key | SKJJIFRWCCSXGL-UHFFFAOYSA-N |
Synonym | o-3m3fbs, 2,4,6-trimethyl-n-2-trifluoromethyl phenyl benzenesulfonamide, tocris-1942, benzenesulfonamide,2,4,6-trimethyl-n-2-trifluoromethyl phenyl |
PubChem CID | 770820 |
IUPAC Name | 2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide |
SMILES | CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15450437
|
Thermo Scientific Alfa Aesar
J65391.MA |
10 mg |
£83.10
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15460437
|
Thermo Scientific Alfa Aesar
J65391.MF |
50 mg | |||||||
Description
An inactive analog of m-3M3FBS. Used as a negative control.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- An inactive analog of m-3M3FBS
- Used as a negative control
Chemical Identifiers
313981-55-4 | |
343.36 | |
SKJJIFRWCCSXGL-UHFFFAOYSA-N | |
770820 | |
CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F |
C16H16F3NO2S | |
MFCD00784362 | |
o-3m3fbs, 2,4,6-trimethyl-n-2-trifluoromethyl phenyl benzenesulfonamide, tocris-1942, benzenesulfonamide,2,4,6-trimethyl-n-2-trifluoromethyl phenyl | |
2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide |
Specifications
313981-55-4 | |
C16H16F3NO2S | |
o-3m3fbs, 2,4,6-trimethyl-n-2-trifluoromethyl phenyl benzenesulfonamide, tocris-1942, benzenesulfonamide,2,4,6-trimethyl-n-2-trifluoromethyl phenyl | |
CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F | |
343.36 | |
343.36 | |
2,4,6-Trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide |
White | |
MFCD00784362 | |
SKJJIFRWCCSXGL-UHFFFAOYSA-N | |
2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide | |
770820 | |
Powder |
Safety and Handling
P261-P280a-P305+P351+P338-P304+P340-P405-P501a
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only