missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylaniline, 97%, Thermo Scientific Chemicals

Product Code. 11955211
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-05-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007740
InChI Key KWVPRPSXBZNOHS-UHFFFAOYSA-N
Synonym mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino
PubChem CID 6913
ChEBI CHEBI:82545
IUPAC Name 2,4,6-trimethylaniline
SMILES CC1=CC(=C(C(=C1)C)N)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -5.0°C
Density 0.9600g/mL
Boiling Point 233.0°C
Flash Point 96°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3C6H2NH2
Refractive Index 1.5505 to 1.5525
Quantity 2.5 kg
Beilstein 12,116
Specific Gravity 0.96
Solubility Information Solubility in water: immiscible with water. Other solubilities: slightly soluble in carbon tetrachloride,soluble in most common solvents
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material 2, 4, 6-Trimethylaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Trimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'