missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethylbenzoic acid, 99%, Thermo Scientific Chemicals
£30.20 - £510.00
Chemical Identifiers
CAS | 480-63-7 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.20 |
MDL Number | MFCD00002481 |
InChI Key | FFFIRKXTFQCCKJ-UHFFFAOYSA-N |
Synonym | mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl |
PubChem CID | 10194 |
ChEBI | CHEBI:64830 |
IUPAC Name | 2,4,6-trimethylbenzoic acid |
SMILES | CC1=CC(C)=C(C(O)=O)C(C)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453784
|
Thermo Scientific Alfa Aesar
A11831.09 |
10 g |
£30.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463784
|
Thermo Scientific Alfa Aesar
A11831.18 |
50 g |
£123.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473784
|
Thermo Scientific Alfa Aesar
A11831.30 |
250 g |
£510.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
480-63-7 | |
164.20 | |
FFFIRKXTFQCCKJ-UHFFFAOYSA-N | |
10194 | |
2,4,6-trimethylbenzoic acid |
C10H12O2 | |
MFCD00002481 | |
mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl | |
CHEBI:64830 | |
CC1=CC(C)=C(C(O)=O)C(C)=C1 |
Specifications
480-63-7 | |
C10H12O2 | |
1866187 | |
FFFIRKXTFQCCKJ-UHFFFAOYSA-N | |
2,4,6-trimethylbenzoic acid | |
10194 | |
164.2 | |
2,4,6-Trimethylbenzoic acid |
153°C to 155°C | |
MFCD00002481 | |
mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl | |
CC1=CC(C)=C(C(O)=O)C(C)=C1 | |
164.20 | |
CHEBI:64830 | |
99% |
Safety and Handling
missing translation for 'einecsNumber' : 207-553-9
missing translation for 'rtecsNumber' : DI0887010
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only