missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzoyl chloride, 98+%, Thermo Scientific Chemicals

Product Code. 11437027
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,4,6-Trimethylbenzoyl chloride is used, in the presence of pyridine, to protect OH groups as mesitoate esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylbenzoyl chloride is used, in the presence of pyridine, to protect OH groups as mesitoate esters.

Solubility
Reacts slowly with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Air and moisture sensitive. Store under nitrogen. Incompatible with strong oxidizing agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-18-1
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD00013650
InChI Key UKRQMDIFLKHCRO-UHFFFAOYSA-N
Synonym mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride
PubChem CID 97038
IUPAC Name 2,4,6-trimethylbenzoyl chloride
SMILES CC1=CC(=C(C(=C1)C)C(=O)Cl)C

Specifications

Boiling Point 143°C to 146°C (60 mmHg)
Flash Point >110°C (230°F)
Quantity 250 g
UN Number UN3265
Beilstein 776108
Formula Weight 182.65
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Trimethylbenzoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'