missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzoyl chloride, 98%, Thermo Scientific Chemicals

£60.20 - £229.00

Chemical Identifiers

CAS 938-18-1
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.65
MDL Number MFCD00013650
InChI Key UKRQMDIFLKHCRO-UHFFFAOYSA-N
Synonym mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride
PubChem CID 97038
IUPAC Name 2,4,6-trimethylbenzoyl chloride
SMILES CC1=CC(=C(C(=C1)C)C(=O)Cl)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10119232
View Documents Promotion Details
Thermo Scientific Acros
244280100
10 g Glass bottle
£60.20
10g
Please to purchase this item. Need a web account? Register with us today!
10522763
View Documents
Thermo Scientific Acros
244280500
50 g Glass bottle
£229.00
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

938-18-1
182.65
UKRQMDIFLKHCRO-UHFFFAOYSA-N
97038
CC1=CC(=C(C(=C1)C)C(=O)Cl)C
C10H11ClO
MFCD00013650
mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride
2,4,6-trimethylbenzoyl chloride

Specifications

938-18-1
100.0
143°C to 146°C (60.0 mmHg)
97.5% min. (GC)
1.5280 to 1.53
mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride
CC1=CC(=C(C(=C1)C)C(=O)Cl)C
182.65
182.65
Liquid
97.5
Yellow
Authentic
C10H11ClO
MFCD00013650
UKRQMDIFLKHCRO-UHFFFAOYSA-N
2,4,6-trimethylbenzoyl chloride
97038
98%
2, 4, 6-Trimethylbenzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 213-339-6

Product Identifier
  • 2,4,6-Trimethylbenzoyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only