missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2,4,6-Trimethylphenylhydrazine hydrochloride is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,6-Trimethylphenylhydrazine hydrochloride is used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
2,4,6-Trimethylphenylhydrazine hydrochloride is used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∽185°C (decomposition) |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 3700289 |
Solubility Information | Soluble in water. |
Formula Weight | 186.69 |
Percent Purity | 97% |
Chemical Name or Material | 2,4,6-Trimethylphenylhydrazine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'