missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Biphenylyl)piperazine, 95%, Thermo Scientific™

£97.90 - £272.00

Chemical Identifiers

CAS 105242-10-2
Molecular Formula C16H18N2
Molecular Weight (g/mol) 238.334
MDL Number MFCD05864686
InChI Key QUINYTMXFWCKTN-UHFFFAOYSA-N
Synonym 2-biphenyl-4-yl-piperazine, 2-4-phenylphenyl piperazine, 2-4-biphenylyl piperazine, 2-biphenyl-4-ylpiperazine, 2-1,1'-biphenyl-4-yl piperazine, pubchem9326, acmc-20a0oz, 2-4-biphenylyl piperazin
PubChem CID 2771747
IUPAC Name 2-(4-phenylphenyl)piperazine
SMILES C1CNC(CN1)C2=CC=C(C=C2)C3=CC=CC=C3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15473547
View Documents
Thermo Scientific Alfa Aesar
H26820.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11376949
View Documents
Thermo Scientific Alfa Aesar
H26820.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

105242-10-2
238.334
QUINYTMXFWCKTN-UHFFFAOYSA-N
2771747
C1CNC(CN1)C2=CC=C(C=C2)C3=CC=CC=C3
C16H18N2
MFCD05864686
2-biphenyl-4-yl-piperazine, 2-4-phenylphenyl piperazine, 2-4-biphenylyl piperazine, 2-biphenyl-4-ylpiperazine, 2-1,1'-biphenyl-4-yl piperazine, pubchem9326, acmc-20a0oz, 2-4-biphenylyl piperazin
2-(4-phenylphenyl)piperazine

Specifications

105242-10-2
C16H18N2
Air Sensitive
QUINYTMXFWCKTN-UHFFFAOYSA-N
2-(4-phenylphenyl)piperazine
2771747
95%
156°C to 159°C
MFCD05864686
2-biphenyl-4-yl-piperazine, 2-4-phenylphenyl piperazine, 2-4-biphenylyl piperazine, 2-biphenyl-4-ylpiperazine, 2-1,1'-biphenyl-4-yl piperazine, pubchem9326, acmc-20a0oz, 2-4-biphenylyl piperazin
C1CNC(CN1)C2=CC=C(C=C2)C3=CC=CC=C3
238.334
238.34
2-(4-Biphenylyl)piperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications