missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Bromo-1H-pyrazol-1-yl)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 1174064-66-4 |
Assay Percent Range | 97% |
Molecular Formula | C10H7BrN2O |
MDL Number | MFCD12756233 |
Quantity | 1 g |
Synonym | 2-4-bromo-1h-pyrazol-1-yl benzaldehyde, 2-4-bromopyrazol-1-yl benzaldehyde, 2-4-bromopyrazolyl benzaldehyde |
InChI Key | SHWJJFKWHUIAFC-UHFFFAOYSA-N |
SMILES | BrC1=CN(N=C1)C1=CC=CC=C1C=O |
IUPAC Name | 2-(4-bromopyrazol-1-yl)benzaldehyde |
Molecular Weight (g/mol) | 251.08 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'