Learn More
2-(4-Bromophenoxy)-N,N-dimethylethylamine, 97%, Thermo Scientific™
£26.92 - £311.00
Chemical Identifiers
CAS | 2474-07-9 |
---|---|
Molecular Formula | C10H14BrNO |
Molecular Weight (g/mol) | 244.13 |
MDL Number | MFCD00274471 |
InChI Key | MOVOYJFCKMYLHQ-UHFFFAOYSA-N |
Synonym | 2-4-bromophenoxy-n,n-dimethylethanamine, 2-4-bromophenoxy-n,n-dimethylethylamine, 2-4-bromophenoxy ethyl dimethylamine, 4-2-n,n-dimethylethoxy phenylbromide, 4-2-n,n-dimethylaminoethoxy phenylbromide, 2-4-bromo-phenoxy-ethyl-dimethyl-amine, ethanamine, 2-4-bromophenoxy-n,n-dimethyl, n-2-4-bromophenoxy ethyl-n,n-dimethylamine, 2-4-bromo-phenoxy-ethyl-dimethyl, 4-2-dimethylaminoethoxy bromobenzene |
PubChem CID | 213877 |
IUPAC Name | 2-(4-bromophenoxy)-N,N-dimethylethanamine |
SMILES | CN(C)CCOC1=CC=C(C=C1)Br |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsAn intermediate for the synthesis of Tamoxifen.
Solubility
Soluble in chloroform, dichloromethane.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
2474-07-9 | |
244.13 | |
MOVOYJFCKMYLHQ-UHFFFAOYSA-N | |
213877 | |
CN(C)CCOC1=CC=C(C=C1)Br |
C10H14BrNO | |
MFCD00274471 | |
2-4-bromophenoxy-n,n-dimethylethanamine, 2-4-bromophenoxy-n,n-dimethylethylamine, 2-4-bromophenoxy ethyl dimethylamine, 4-2-n,n-dimethylethoxy phenylbromide, 4-2-n,n-dimethylaminoethoxy phenylbromide, 2-4-bromo-phenoxy-ethyl-dimethyl-amine, ethanamine, 2-4-bromophenoxy-n,n-dimethyl, n-2-4-bromophenoxy ethyl-n,n-dimethylamine, 2-4-bromo-phenoxy-ethyl-dimethyl, 4-2-dimethylaminoethoxy bromobenzene | |
2-(4-bromophenoxy)-N,N-dimethylethanamine |
Specifications
2474-07-9 | |
MFCD00274471 | |
2-4-bromophenoxy-n,n-dimethylethanamine, 2-4-bromophenoxy-n,n-dimethylethylamine, 2-4-bromophenoxy ethyl dimethylamine, 4-2-n,n-dimethylethoxy phenylbromide, 4-2-n,n-dimethylaminoethoxy phenylbromide, 2-4-bromo-phenoxy-ethyl-dimethyl-amine, ethanamine, 2-4-bromophenoxy-n,n-dimethyl, n-2-4-bromophenoxy ethyl-n,n-dimethylamine, 2-4-bromo-phenoxy-ethyl-dimethyl, 4-2-dimethylaminoethoxy bromobenzene | |
MOVOYJFCKMYLHQ-UHFFFAOYSA-N | |
2-(4-bromophenoxy)-N,N-dimethylethanamine | |
213877 | |
97% |
C10H14BrNO | |
UN2735 | |
Soluble in chloroform,dichloromethane. | |
CN(C)CCOC1=CC=C(C=C1)Br | |
244.13 | |
244.13 | |
2-(4-Bromophenoxy)-N,N-dimethylethylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures