missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole, 98+%, Thermo Scientific™

Product Code. 11352666
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21510-43-0
Molecular Formula C14H9BrN2O
Molecular Weight (g/mol) 301.14
MDL Number MFCD00050997
InChI Key CNLVYZSUMYQALH-UHFFFAOYSA-N
PubChem CID 88931
IUPAC Name 2-(4-bromophenyl)-5-phenyl-1,3,4-oxadiazole
SMILES BrC1=CC=C(C=C1)C1=NN=C(O1)C1=CC=CC=C1

Specifications

Melting Point 167°C to 172°C
Quantity 5 g
Beilstein 204891
Synonym 2-4-bromophenyl-5-phenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-bromophenyl-5-phenyl, 5-4-bromophenyl-2-phenyl-1,3,4-oxadiazole, acmc-1cq4f, cambridge id 5570574, 1,3,4-oxadiazole,2-4-bromophenyl-5-phenyl, 2-4-bromo-phenyl-5-phenyl-1,3,4 oxadiazo le
Formula Weight 301.14
Percent Purity ≥98%
Chemical Name or Material 2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'