missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine, 95%, Thermo Scientific™

Product Code. 10421704
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 613239-76-2
Molecular Formula C13H9ClF3N
Molecular Weight (g/mol) 271.67
MDL Number MFCD09817479
InChI Key JTXHHQDYEHXWPC-UHFFFAOYSA-N
Synonym 2-4-chloromethyl phenyl-5-trifluoromethyl pyridine, 4-5-trifluoromethyl pyridin-2-yl benzyl chloride, 2-4-chloromethyl-phenyl 5-trifluoromethyl-pyridine, 2-4-chloromethyl-phenyl-5-trifluoromethyl-pyridine
PubChem CID 22280067
IUPAC Name 2-[4-(chloromethyl)phenyl]-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CN=C(C=C1)C1=CC=C(CCl)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 271.67
Percent Purity 95%
Chemical Name or Material 2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.