missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)-2-methylmorpholine, 99%, Thermo Scientific Chemicals

Product Code. 11489800
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109461-44-1
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.689
MDL Number MFCD08061115
InChI Key IJDDASQRAPIORY-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl-2-methylmorpholine, 2-4-chloro-phenyl-2-methyl-morpholine, acmc-1c8s6, morpholine,2-4-chlorophenyl-2-methyl
PubChem CID 3066043
IUPAC Name 2-(4-chlorophenyl)-2-methylmorpholine
SMILES CC1(CNCCO1)C2=CC=C(C=C2)Cl

Specifications

Quantity 1 g
Formula Weight 211.69
Percent Purity 99%
Chemical Name or Material 2-(4-Chlorophenyl)-2-methylmorpholine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'