missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Chlorophenyl)malondialdehyde, 95%, Thermo Scientific™
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Specifications
Specifications
CAS | 205676-17-1 |
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 137°C to 143°C |
Color | Beige or White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass bottle |
Molecular Formula | C9H7ClO2 |
MDL Number | MFCD00216505 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0223531 | Certificate of Analysis | 30/05/2022 | 425200025 |
Safety and Handling


Product Identifier
- 2-(4-Chlorophenyl)malondialdehyde
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-(4-Chlorophenyl)malondialdehyde, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction